Assay Details
Gene Symbol: Pank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106373.3 | NP_001099843.1 | 6 - 7 | 1234 | 73 | |
XM_006231276.2 | XP_006231338.1 | 6 - 7 | 4537 | 73 | ||
XM_006231277.2 | XP_006231339.1 | 5 - 6 | 4365 | 73 | ||
XM_006231278.2 | XP_006231340.1 | 7 - 8 | 1885 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P592790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43664 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P592790 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P592790 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P592790 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18824 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P592790 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P173876 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87707 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47060 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34192 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70803 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93891 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79035 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10729807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729808 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729803 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729806 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10729802 | - | - |
Rat Genome 230 2.0 Array | 1382924_at | - | - |
Rat Genome 230 2.0 Array | 1382924_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382924_at | - | - |
Affymetrix HT Rat Focus Array | 1382924_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100061067.rn.1 | TC0100007603.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100061068.rn.1 | TC0100007603.rn.1 | RTA TC |
Rat Expression Set 230 | 1382924_PM_at | - | - |
Rat Expression Set 230 | 1382924_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729802 | - | - |
Rat Genome U34 Set | rc_AA850195_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22072471 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356675 | - | - |
