Assay Details
Gene Symbol: Eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004223.2 | NP_001004223.1 | 8 - 9 | 1159 | 133 | |
GenBank mRNA | BC079398.1 | - | 9 - 10 | 1168 | 133 | |
BC098895.1 | - | 8 - 9 | 1159 | 133 | ||
FQ212596.1 | - | 9 - 10 | 1177 | 133 | ||
FQ213179.1 | - | 9 - 10 | 1176 | 133 | ||
FQ214210.1 | - | 9 - 10 | 1174 | 133 | ||
FQ215853.1 | - | 9 - 10 | 1205 | 133 | ||
FQ218274.1 | - | 9 - 10 | 1175 | 133 | ||
FQ219298.1 | - | 9 - 10 | 1177 | 133 | ||
FQ222930.1 | - | 9 - 10 | 1176 | 133 | ||
FQ223614.1 | - | 9 - 10 | 1175 | 133 | ||
FQ224956.1 | - | 8 - 9 | 1135 | 133 | ||
FQ226853.1 | - | 6 - 7 | 848 | 133 | ||
FQ228703.1 | - | 9 - 10 | 1175 | 133 | ||
FQ228918.1 | - | 9 - 10 | 1177 | 133 | ||
FQ230995.1 | - | 9 - 10 | 1175 | 133 | ||
FQ231923.1 | - | 9 - 10 | 1177 | 133 | ||
FQ233868.1 | - | 9 - 10 | 1175 | 133 | ||
FQ234035.1 | - | 9 - 10 | 1175 | 133 | ||
FQ235034.1 | - | 9 - 10 | 1327 | 133 |
Gene Symbol: Slamf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250250.2 | XP_006250312.1 | 7 | 3446 | 133 | |
GenBank mRNA | FQ230857.1 | - | 3 | 2380 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
