Assay Details
Gene Symbol: Ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006972.1 | NP_001006973.1 | 4 - 5 | 518 | 97 | |
GenBank mRNA | BC083709.1 | - | 4 - 5 | 518 | 97 | |
FQ214688.1 | - | 4 - 5 | 525 | 97 | ||
FQ217076.1 | - | 4 - 5 | 575 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
