Assay Details
Gene Symbol: Echs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078623.2 | NP_511178.1 | 3 - 4 | 435 | 98 | |
GenBank mRNA | BC064655.1 | - | 3 - 4 | 435 | 98 | |
FQ218439.1 | - | 2 - 3 | 294 | 98 | ||
X15958.1 | - | 3 - 4 | 429 | 98 |
Gene Symbol: LOC100911186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748886.2 | XP_003748934.1 | 3 - 4 | 463 | 98 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
