Assay Details
Gene Symbol: Echs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078623.2 | NP_511178.1 | 7 - 8 | 824 | 120 | |
GenBank mRNA | BC064655.1 | - | 7 - 8 | 824 | 120 | |
X15958.1 | - | 7 - 8 | 818 | 120 |
Gene Symbol: LOC100911186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748886.2 | XP_003748934.1 | 7 - 8 | 852 | 120 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
