Assay Details
Gene Symbol: Trpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106331.1 | NP_001099801.1 | 4 - 5 | 819 | 112 | |
XM_006230761.2 | XP_006230823.1 | 4 - 5 | 509 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
