Assay Details
Gene Symbol: Rcor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013994.1 | NP_001014016.1 | 4 - 5 | 670 | 66 | |
XM_008760113.1 | XP_008758335.1 | 5 - 6 | 520 | 66 | ||
XM_008760114.1 | XP_008758336.1 | 5 - 6 | 587 | 66 | ||
XM_008760115.1 | XP_008758337.1 | 5 - 6 | 706 | 66 | ||
GenBank mRNA | BC089209.1 | - | 4 - 5 | 670 | 66 |
Gene Symbol: LOC102552640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770611.1 | XP_008768833.1 | 4 - 5 | 664 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
