Assay Details
Gene Symbol: Rcor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013994.1 | NP_001014016.1 | 6 - 7 | 958 | 57 | |
XM_008760113.1 | XP_008758335.1 | 7 - 8 | 808 | 57 | ||
XM_008760114.1 | XP_008758336.1 | 7 - 8 | 875 | 57 | ||
XM_008760115.1 | XP_008758337.1 | 7 - 8 | 994 | 57 | ||
GenBank mRNA | BC089209.1 | - | 6 - 7 | 958 | 57 |
Gene Symbol: LOC102552640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770611.1 | XP_008768833.1 | 6 - 7 | 952 | 57 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
