Assay Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181627.2 | NP_853658.1 | 118 | 130 | ||
GenBank mRNA | AY291062.1 | - | 118 | 130 | ||
AY919342.1 | - | 118 | 130 | |||
FJ517605.1 | - | 92 | 130 | |||
FJ517607.1 | - | 1 - 2 | 92 | 130 | ||
FQ213378.1 | - | 135 | 130 | |||
FQ213584.1 | - | 135 | 130 | |||
FQ214037.1 | - | 137 | 129 | |||
FQ227907.1 | - | 135 | 130 | |||
FQ229190.1 | - | 135 | 130 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY291062.1, AY919342.1, FJ517605.1, FJ517607.1, NM_181627.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
