Assay Details
Gene Symbol: Asic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022227.1 | NP_071563.1 | 1 - 2 | 120 | 124 | |
GenBank mRNA | Y19034.1 | - | 1 - 2 | 120 | 124 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_022227.1, Y19034.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
