Assay Details
Gene Symbol: mrpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007637.1 | NP_001007638.1 | 2 - 3 | 267 | 84 | |
XM_006232604.2 | XP_006232666.1 | 2 - 3 | 559 | 84 | ||
XM_006232605.2 | XP_006232667.1 | 2 - 3 | 562 | 84 | ||
XM_006232607.2 | XP_006232669.1 | 2 - 3 | 449 | 84 | ||
XM_006232608.2 | XP_006232670.1 | 2 - 3 | 479 | 84 | ||
GenBank mRNA | BC082018.1 | - | 2 - 3 | 267 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
