Assay Details
Gene Symbol: mrpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007637.1 | NP_001007638.1 | 4 - 5 | 468 | 124 | |
XM_006232604.2 | XP_006232666.1 | 4 - 5 | 760 | 124 | ||
XM_006232605.2 | XP_006232667.1 | 4 - 5 | 763 | 124 | ||
XM_006232607.2 | XP_006232669.1 | 4 - 5 | 650 | 124 | ||
XM_006232608.2 | XP_006232670.1 | 4 - 5 | 680 | 124 | ||
GenBank mRNA | BC082018.1 | - | 4 - 5 | 468 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
