Assay Details
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004198.1 | NP_001004198.1 | 5 - 6 | 879 | 124 | |
XM_006227980.2 | XP_006228042.1 | 5 - 6 | 822 | 124 | ||
XM_008758747.1 | XP_008756969.1 | 5 - 6 | 822 | 124 | ||
XM_008758748.1 | XP_008756970.1 | 5 - 6 | 1030 | 124 | ||
GenBank mRNA | BC081939.1 | - | 5 - 6 | 879 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
