Assay Details
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134553.1 | NP_001128025.1 | 15 - 16 | 3864 | 76 | |
XM_006236306.2 | XP_006236368.1 | 15 - 16 | 4953 | 76 | ||
XM_006236307.2 | XP_006236369.1 | 14 - 15 | 4902 | 76 | ||
XM_006236308.2 | XP_006236370.1 | 14 - 15 | 4896 | 76 | ||
GenBank mRNA | FQ212455.1 | - | 1 - 2 | 3436 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
