Assay Details
Gene Symbol: Zc3hav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173045.2 | NP_766633.2 | 5 - 6 | 2120 | 77 | |
XM_006236295.2 | XP_006236357.1 | 5 - 6 | 2181 | 77 | ||
GenBank mRNA | AF521008.1 | - | 5 - 6 | 1804 | 77 | |
BC131842.1 | - | 5 - 6 | 2120 | 77 | ||
FQ230840.1 | - | 5 - 6 | 2143 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
