Assay Details
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034107.1 | NP_001029279.1 | 1 - 2 | 98 | 68 | |
XM_006236140.2 | XP_006236202.1 | 1 - 2 | 173 | 68 | ||
GenBank mRNA | BC100082.1 | - | 1 - 2 | 98 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC100082.1, NM_001034107.1, XM_006236140.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
