Assay Details
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044238.1 | NP_001037703.1 | 8 - 9 | 880 | 144 | |
GenBank mRNA | BC105620.1 | - | 8 - 9 | 880 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1007438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137633 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1007438 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1007438 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1007438 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1007438 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84467 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6704 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10746013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746010 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746002 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10746000 | - | - |
Rat Genome 230 2.0 Array | 1378235_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385302_at | - | - |
Rat Genome 230 2.0 Array | 1378235_at | - | - |
Rat Genome 230 2.0 Array | 1385302_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385302_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385302_at | - | - |
Affymetrix HT Rat Focus Array | 1378235_at | - | - |
Affymetrix HT Rat Focus Array | 1378235_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000031213.rn.1 | TC1000003478.rn.1 | RTA TC |
Rat Expression Set 230 | 1385302_PM_at | - | - |
Rat Expression Set 230 | 1385302_at | - | - |
Rat Expression Set 230 | 1378235_PM_at | - | - |
Rat Expression Set 230 | 1378235_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746000 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369438 | - | - |
