Assay Details
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168612.1 | NP_001162083.1 | 147 | 111 | ||
GenBank mRNA | CB611803.1 | - | 147 | 111 | ||
CO555844.1 | - | 146 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
