Assay Details
Gene Symbol: Rpl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191589.1 | NP_001178518.1 | 3 - 4 | 401 | 80 | |
GenBank mRNA | FQ214717.1 | - | 3 - 4 | 420 | 80 | |
FQ215051.1 | - | 3 - 4 | 383 | 80 | ||
FQ216207.1 | - | 3 - 4 | 383 | 80 | ||
FQ216222.1 | - | 3 - 4 | 383 | 80 | ||
FQ216295.1 | - | 3 - 4 | 395 | 80 | ||
FQ216388.1 | - | 3 - 4 | 421 | 80 | ||
FQ216791.1 | - | 3 - 4 | 420 | 80 | ||
FQ224678.1 | - | 3 - 4 | 429 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
