Assay Details
Gene Symbol: Rpl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191589.1 | NP_001178518.1 | 8 - 9 | 1077 | 66 | |
GenBank mRNA | FQ214717.1 | - | 8 - 9 | 1096 | 66 | |
FQ215051.1 | - | 8 - 9 | 1059 | 66 | ||
FQ216207.1 | - | 8 - 9 | 1059 | 66 | ||
FQ216222.1 | - | 8 - 9 | 1059 | 66 | ||
FQ216295.1 | - | 8 - 9 | 1071 | 66 | ||
FQ216388.1 | - | 8 - 9 | 1097 | 66 | ||
FQ216791.1 | - | 8 - 9 | 1097 | 66 | ||
FQ224678.1 | - | 8 - 9 | 1106 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
