Assay Details
Gene Symbol: Rpl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191589.1 | NP_001178518.1 | 9 - 10 | 1194 | 111 | |
GenBank mRNA | FQ216222.1 | - | 9 - 10 | 1176 | 111 | |
FQ216645.1 | - | 9 - 10 | 1175 | 111 | ||
FQ216791.1 | - | 9 - 10 | 1214 | 111 | ||
FQ224678.1 | - | 9 - 10 | 1223 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
