Assay Details
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109035.1 | NP_001102505.1 | 4 - 5 | 662 | 66 | |
XM_006246786.2 | XP_006246848.1 | 5 - 6 | 1251 | 66 | ||
XM_006246789.2 | XP_006246851.1 | 5 - 6 | 1252 | 66 | ||
XM_008767857.1 | XP_008766079.1 | 5 - 6 | 1252 | 66 | ||
XM_008767858.1 | XP_008766080.1 | 5 - 6 | 1252 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
