Assay Details
Gene Symbol: Sepsecs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128287.2 | NP_001121759.1 | 2 - 3 | 272 | 75 | |
GenBank mRNA | BC166584.1 | - | 2 - 3 | 272 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
