Assay Details
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106631.1 | NP_001100101.1 | 7 - 8 | 658 | 112 | |
XM_008770190.1 | XP_008768412.1 | 6 - 7 | 1329 | 112 | ||
XM_008770191.1 | XP_008768413.1 | 6 - 7 | 570 | 112 | ||
XM_008770192.1 | XP_008768414.1 | 6 - 7 | 2748 | 112 | ||
XM_008770193.1 | XP_008768415.1 | 7 - 8 | 741 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
