Assay Details
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106631.1 | NP_001100101.1 | 10 - 11 | 1048 | 151 | |
XM_008770190.1 | XP_008768412.1 | 9 - 10 | 1719 | 151 | ||
XM_008770191.1 | XP_008768413.1 | 9 - 10 | 960 | 151 | ||
XM_008770192.1 | XP_008768414.1 | 9 - 10 | 3138 | 151 | ||
XM_008770193.1 | XP_008768415.1 | 10 - 11 | 1131 | 151 |
Gene Symbol: Rbpjl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237737.2 | XP_006237799.1 | 2 | 970 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
