Assay Details
Gene Symbol: Lamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017068.2 | NP_058764.2 | 5 - 6 | 853 | 82 | |
XM_006257486.2 | XP_006257548.1 | 5 - 6 | 884 | 82 | ||
GenBank mRNA | BC061990.1 | - | 5 - 6 | 853 | 82 | |
D90211.1 | - | 5 - 6 | 837 | 82 | ||
M32016.1 | - | 4 - 5 | 691 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
