Assay Details
Gene Symbol: Lamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017068.2 | NP_058764.2 | 6 - 7 | 974 | 162 | |
XM_006257486.2 | XP_006257548.1 | 6 - 7 | 1005 | 162 | ||
GenBank mRNA | BC061990.1 | - | 6 - 7 | 974 | 162 | |
D90211.1 | - | 6 - 7 | 958 | 162 | ||
FQ211284.1 | - | 6 - 7 | 975 | 162 | ||
M32016.1 | - | 5 - 6 | 812 | 162 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
