Assay Details
Gene Symbol: Xiap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022231.2 | NP_071567.1 | 5 - 6 | 1383 | 177 | |
XM_006257506.2 | XP_006257568.1 | 4 - 5 | 1209 | 177 | ||
XM_006257508.2 | XP_006257570.1 | 4 - 5 | 1323 | 177 | ||
XM_006257509.2 | XP_006257571.1 | 4 - 5 | 1224 | 177 | ||
XM_008773529.1 | XP_008771751.1 | 5 - 6 | 1350 | 177 | ||
GenBank mRNA | AB033366.1 | - | 5 - 6 | 1382 | 177 | |
AF183429.1 | - | 3 - 4 | 1053 | 177 | ||
AF304334.1 | - | 1978 | 177 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
