Assay Details
Gene Symbol: Xiap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022231.2 | NP_071567.1 | 2 - 3 | 295 | 115 | |
XM_008773529.1 | XP_008771751.1 | 2 - 3 | 262 | 115 | ||
GenBank mRNA | AB033366.1 | - | 2 - 3 | 294 | 115 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB033366.1, NM_022231.2, XM_008773529.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
