Assay Details
Gene Symbol: Stk26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191736.1 | NP_001178665.1 | 8 - 9 | 1150 | 79 | |
XM_006257573.2 | XP_006257635.1 | 10 - 11 | 1637 | 79 | ||
XM_008773567.1 | XP_008771789.1 | 9 - 10 | 2349 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
