Assay Details
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145789.2 | NP_665732.2 | 7 - 8 | 922 | 149 | |
GenBank mRNA | AY044251.1 | - | 7 - 8 | 906 | 149 | |
BC093615.1 | - | 7 - 8 | 922 | 149 |
Gene Symbol: LOC100360218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751077.3 | XP_003751125.1 | 1 | 948 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
