Assay Details
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145789.2 | NP_665732.2 | 8 - 9 | 1052 | 153 | |
GenBank mRNA | AY044251.1 | - | 8 - 9 | 1036 | 153 | |
BC093615.1 | - | 8 - 9 | 1052 | 153 |
Gene Symbol: LOC100360218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751077.3 | XP_003751125.1 | 1 | 1078 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
