Assay Details
Gene Symbol: Mbnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106949.1 | NP_001100419.1 | 5 - 6 | 995 | 62 | |
XM_008773559.1 | XP_008771781.1 | 5 - 6 | 1597 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
