Assay Details
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038588.1 | NP_001033677.1 | 2 - 3 | 381 | 65 | |
GenBank mRNA | AF222852.1 | - | 2 - 3 | 371 | 65 | |
BC158806.1 | - | 2 - 3 | 381 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
