Assay Details
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229043.1 | XP_006229105.1 | 1 - 2 | 111 | 74 | |
GenBank mRNA | FQ229080.1 | - | 1 - 2 | 45 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229043.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
