Assay Details
Gene Symbol: Rab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017355.1 | NP_059051.1 | 2 - 3 | 179 | 79 | |
GenBank mRNA | FQ227674.1 | - | 2 - 3 | 211 | 79 | |
X78605.1 | - | 2 - 3 | 179 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
