Assay Details
Gene Symbol: Bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012782.1 | NP_036914.1 | 3 - 4 | 383 | 80 | |
GenBank mRNA | BC161819.1 | - | 3 - 4 | 388 | 80 | |
CB579195.1 | - | 3 - 4 | 389 | 80 | ||
J02827.1 | - | 3 - 4 | 367 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
