Assay Details
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ212221.1 | - | 12 - 13 | 1575 | 117 | |
FQ213451.1 | - | 12 - 13 | 1627 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
