Assay Details
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100802.1 | NP_001094272.1 | 16 - 17 | 1882 | 106 | |
XM_008759179.1 | XP_008757401.1 | 15 - 16 | 1763 | 106 | ||
GenBank mRNA | BC161904.1 | - | 16 - 17 | 1882 | 106 | |
DQ462463.1 | - | 1 - 2 | 104 | 106 | ||
FQ212221.1 | - | 16 - 17 | 1872 | 106 | ||
FQ213451.1 | - | 16 - 17 | 1924 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
