Assay Details
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100802.1 | NP_001094272.1 | 5 - 6 | 719 | 66 | |
XM_008759179.1 | XP_008757401.1 | 4 - 5 | 567 | 66 | ||
GenBank mRNA | BC161904.1 | - | 5 - 6 | 719 | 66 | |
FQ212221.1 | - | 5 - 6 | 679 | 66 | ||
FQ213451.1 | - | 5 - 6 | 731 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
