Assay Details
Gene Symbol: Cblc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034920.2 | NP_001030092.1 | 11 - 12 | 1584 | 74 | |
GenBank mRNA | BC105776.1 | - | 11 - 12 | 1569 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
