Assay Details
Gene Symbol: Cblc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034920.2 | NP_001030092.1 | 4 - 5 | 804 | 62 | |
GenBank mRNA | BC105776.1 | - | 4 - 5 | 789 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
