Assay Details
Gene Symbol: Ifngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108313.1 | NP_001101783.1 | 5 - 6 | 812 | 95 | |
GenBank mRNA | FQ227918.1 | - | 1 - 2 | 102 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
