Assay Details
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248041.2 | XP_006248103.1 | 1 - 2 | 348 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
