Assay Details
Gene Symbol: Dcaf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173558.1 | NP_001167029.1 | 1 - 2 | 137 | 118 | |
XM_006241605.2 | XP_006241667.1 | 1 - 2 | 96 | 118 | ||
GenBank mRNA | EV763930.1 | - | 1 - 2 | 137 | 118 | |
FQ228874.1 | - | 1 - 2 | 147 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
