Assay Details
Gene Symbol: Dcaf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173558.1 | NP_001167029.1 | 8 - 9 | 1012 | 97 | |
XM_006241605.2 | XP_006241667.1 | 8 - 9 | 992 | 97 | ||
GenBank mRNA | BC158730.1 | - | 6 - 7 | 606 | 97 | |
FQ228874.1 | - | 8 - 9 | 1021 | 97 | ||
FQ229670.1 | - | 6 - 7 | 671 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
