Assay Details
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135876.1 | NP_001129348.1 | 5 - 6 | 722 | 143 | |
XM_006242164.2 | XP_006242226.1 | 5 - 6 | 735 | 143 | ||
XM_008765766.1 | XP_008763988.1 | 4 - 5 | 432 | 143 | ||
GenBank mRNA | BC161991.1 | - | 5 - 6 | 694 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
