Assay Details
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134883.1 | NP_001128355.1 | 1 - 2 | 164 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P140591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48685 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P140591 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P140591 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P140591 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P743637 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P140591 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10903507 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903510 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903509 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903505 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903508 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903506 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903501 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903503 | - | - |
Rat Genome 230 2.0 Array | 1374108_at | - | - |
Rat Genome 230 2.0 Array | 1374108_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374108_at | - | - |
Affymetrix HT Rat Focus Array | 1374108_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700023598.rn.1 | TC0700003048.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023597.rn.1 | TC0700003048.rn.1 | RTA TC |
Rat Expression Set 230 | 1374108_at | - | - |
Rat Expression Set 230 | 1374108_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903503 | - | - |
Rat Genome U34 Set | rc_AI012317_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22035701 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350073 | - | - |
