Assay Details
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164060.1 | NP_001157532.1 | 5 - 6 | 1013 | 90 | |
NM_053517.2 | NP_445969.2 | 6 - 7 | 580 | 90 | ||
GenBank mRNA | BC088298.1 | - | 6 - 7 | 580 | 90 | |
D83015.1 | - | 5 - 6 | 361 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
