Assay Details
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001059906.5 | XP_001059906.3 | 1 - 2 | 1133 | 90 | |
GenBank mRNA | BC169031.1 | - | 1 - 2 | 191 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
